Tetramethylammonium hydrogen sulfate ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetramethylammonium bisulfate

T0840-25G
TCT0840-025GEA 116.6 USD
TCT0840-025G
Tetramethylammonium hydrogen sulfate ≥96.0% (by titrimetric analysis)
Tetramethylammonium hydrogen sulphate

CAS Number: 80526-82-5
MDL Number: MFCD00036149
Molecular Formula: C4H13NO4S
Molecular Weight: 171.21
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Color: White

Formula: C₄H₁₃NO₄S
MW: 171.22 g/mol
MDL Number: MFCD00036149
CAS Number: 80526-82-5
EINECS: 279-490-5
https://us.vwr.com/stibo/web/std.lang.all/49/51/28994951.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR