TCIAT0818

T0818-5G T0818-25G
TCT0818-025GEA 365.7 USD
TCT0818-025G TCT0818-005G
TCIAT0818
CAS Number: 940-69-2
MDL Number: MFCD00005475
Molecular Formula: C8H15NOS2
Molecular Weight: 205.33
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 130

Other Product Names / Synonyms: 1,2-Dithiolane-3-valeroylamide, DL-6,8-Epidithiooctanamide, DL-alpha-Lipoamide
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR