TCIAT0752

T0752-5G
TCT0752-005GEA 447.52 USD
TCT0752-005G
TCIAT0752
CAS Number: 23677-93-2
MDL Number: MFCD00042511
Molecular Formula: C10H8CuF6O4
Molecular Weight: 369.71
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Pale Blue
Melting point (°C): 195

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Copper(II) Trifluoroacetylacetonate, Trifluoroacetylacetono Copper(II) Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR