2-Allylcyclohexanone ≥97.0%

Supplier: TCI America

A2230-5G A2230-25G
TCA2230-25GEA 394.9 USD
TCA2230-25G TCA2230-5G
2-Allylcyclohexanone ≥97.0%
2-Allylcyclohexanone

CAS Number: 94-66-6
MDL Number: MFCD00043548
Molecular Formula: C9H14O
Molecular Weight: 138.21
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 109
Specific Gravity (20/20): 0.94

https://us.vwr.com/stibo/web/std.lang.all/04/15/28930415.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR