Tetramethyl-p-benzoquinone ≥98.0%

Supplier: TCI America

Synonyms: Duroquinone2,3,5,6-Tetramethylcyclohexa-2,5-diene-1,4-dioneTetramethylquinoneTetramethyl-1,4-benzoquinone2,3,5,6-Tetramethyl-1,4-benzoquinone

T0672-1G T0672-5G T0672-25G
TCT0672-001GEA 105.11 USD
TCT0672-001G TCT0672-005G TCT0672-25G
Tetramethyl-p-benzoquinone ≥98.0%
Tetramethyl-p-benzoquinone

CAS Number: 527-17-3
MDL Number: MFCD00001604
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 112


Other Product Names / Synonyms: Duroquinone

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 112 °C
MDL Number: MFCD00001604
CAS Number: 527-17-3
EINECS: 208-409-8
Merck Index: 13,03503
https://us.vwr.com/stibo/web/std.lang.all/48/41/28994841.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR