(±)-2-Tetradecanol ≥95.0%

Supplier: TCI America

Synonyms: (±)-2-Hydroxy tetradecane2-Tetradecyl alcohol2-Tetradecanol

T0614-10G
TCT0614-010GEA 243.52 USD
TCT0614-010G
(±)-2-Tetradecanol ≥95.0%
(±)-2-Tetradecanol

CAS Number: 4706-81-4
MDL Number: MFCD00004552
Molecular Formula: C14H30O
Molecular Weight: 214.39
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 170
Melting point (°C): 34
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Tetradecyl Alcohol

Formula: C₁₄H₃₀O
MW: 214.39 g/mol
Boiling Pt: 170 °C
Melting Pt: 34 °C
Density: 0.8315 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004552
CAS Number: 4706-81-4
EINECS: 225-192-5
https://us.vwr.com/stibo/web/std.lang.all/48/09/28994809.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR