2,4,4'-Trihhydroxybenzophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,4,4-Trihydroxybenzophenone2,4,4'-Trihydroxybenzophenone

T0584-10G
TCT0584-010GEA 154 USD
TCT0584-010G
2,4,4'-Trihhydroxybenzophenone ≥98.0% (by HPLC, titration analysis)
2,4,4'-Trihhydroxybenzophenone

CAS Number: 1470-79-7
MDL Number: MFCD00002356
Molecular Formula: C13H10O4
Molecular Weight: 230.22
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 201

Formula: C₁₃H₁₀O₄
MW: 230.22 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: 201 °C
Density: 1.054 g/cm³ (20 °C)
MDL Number: MFCD00002356
CAS Number: 1470-79-7
EINECS: 216-004-2
https://us.vwr.com/stibo/web/std.lang.all/47/88/28994788.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR