1,3,5-Trithiane ≥98.0%
Supplier: TCI America
T0532-25G
TCT0532-025GEA
218
USD
TCT0532-025G
1,3,5-Trithiane ≥98.0%
1,3,5-Trithiane
CAS Number: 291-21-4
MDL Number: MFCD00006653
Molecular Formula: C3H6S3
Molecular Weight: 138.26
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 216
Other Product Names / Synonyms: Thioformaldehyde Trimer, Trithiometaformaldehyde
Formula:
C₃H₆S₃ Melting Pt: 216 °C |
MDL Number:
MFCD00006653 CAS Number: 291-21-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...