3,3',5-Triiodo-L-thyronine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoic acid
T0453-100MG
TCT0453-100MGEA
112.73
USD
TCT0453-100MG
3,3',5-Triiodo-L-thyronine ≥98.0% (by HPLC, titration analysis)
3,3',5-Triiodo-L-thyronine
CAS Number: 6893-02-3
MDL Number: MFCD00002593
Molecular Formula: C15H12I3NO4
Molecular Weight: 650.98
Purity/Analysis Method:
Form: Crystal
Color: White
Specific rotation [a]20/D: 23 deg (C=5, 1mol/L HCl:EtOH=1:2)
Formula:
C₁₅H₁₂I₃NO₄ MW: 650.98 g/mol |
MDL Number:
MFCD00002593 CAS Number: 6893-02-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...