5-Chloro-2-phenoxyaniline ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-2-phenoxyaniline2-Amino-4-chlorodiphenyl ether2-Amino-4-chloro-diphenyl ether

A2222-25G
TCA2222-25GEA 50 USD
TCA2222-25G
5-Chloro-2-phenoxyaniline ≥98.0%
5-Chloro-2-phenoxyaniline

CAS Number: 93-67-4
MDL Number: MFCD00025217
Molecular Formula: C12H10ClNO
Molecular Weight: 219.67
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 43


Other Product Names / Synonyms: 5-Chloro-2-phenoxyaniline

https://us.vwr.com/stibo/web/std.lang.all/04/10/28930410.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR