Bithionol disodium salt ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Bithionol disodium salt
, Bis(3,5-dichloro-2-hydroxyphenyl) sulphide disodium salt
, Bis(3,5-dichloro-2-hydroxyphenyl) sulfide disodium salt
, 2,2'-Thiobis(4,6-dichlorophenol) disodium salt
T0406-25G
TCT0406-025GEA
416.98
USD
TCT0406-025G
Bithionol disodium salt ≥98.0% (by HPLC, titration analysis)
Bithionol disodium salt
CAS Number: 6385-58-6
MDL Number: MFCD00058975
Molecular Formula: C12H6Cl4O2S
Molecular Weight: 400.00
Purity/Analysis Method:
Form: Crystal
Color: Pale Gray
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Bis(3,5-dichloro-2-hydroxyphenyl) Sulfide Disodium Salt, Bithionol Disodium Salt
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...