1,2,3-Trichloropropene ≥95.0%
Supplier: TCI America
T0396-25G
T0396-500G
TCT0396-500GEA
344.09
USD
TCT0396-500G
TCT0396-025G
1,2,3-Trichloropropene ≥95.0%
1,2,3-Trichloropropene
CAS Number: 96-19-5
MDL Number: MFCD00018931
Molecular Formula: C3H3Cl3
Molecular Weight: 145.41
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 82
Specific Gravity (20/20): 1.43
Formula:
C₃H₃Cl₃ MW: 145.41 g/mol Boiling Pt: 145 °C Density: 1.43 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00018931 CAS Number: 96-19-5 EINECS: 202-487-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...