5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulphonamide4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulfonamide

A2219-25G
TCA2219-25GEA 236 USD
TCA2219-25G
5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole ≥98.0% (by HPLC)
5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole

CAS Number: 82212-14-4
Molecular Formula: C7H7ClN6O2S
Molecular Weight: 274.68
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulfonamide

Formula: C₇H₇ClN₆O₂S
MW: 274.68 g/mol
MDL Number: MFCD11870108
CAS Number: 82212-14-4
https://us.vwr.com/stibo/web/std.lang.all/04/07/28930407.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR