m-Toluidine hydrochloride ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
M-toluidine hydrochloride
T0301-25G
TCT0301-025GEA
179
USD
TCT0301-025G
m-Toluidine hydrochloride ≥98.0% (by GC, total nitrogen)
m-Toluidine hydrochloride
CAS Number: 638-03-9
MDL Number: MFCD00070607
Molecular Formula: C7H9N
Molecular Weight: 143.61
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 215
Formula:
C₇H₉N MW: 143.61 g/mol Melting Pt: 215 °C |
MDL Number:
MFCD00070607 CAS Number: 638-03-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...