5,5'-Thiodisalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0208-5G T0208-25G
TCT0208-005GEA 90.3 USD
TCT0208-005G TCT0208-025G
5,5'-Thiodisalicylic acid ≥98.0% (by GC, titration analysis)
5,5'-Thiodisalicylic acid

CAS Number: 1820-99-1
MDL Number: MFCD00020264
Molecular Formula: C14H10O6S
Molecular Weight: 306.29
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White

Formula: C₁₄H₁₀O₆S
MDL Number: MFCD00020264
CAS Number: 1820-99-1
https://us.vwr.com/stibo/web/std.lang.all/45/23/28994523.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR