8-Aminoquinoline-N-oxide ≥98.0% (by GC, titration analysis)

A2211-5G A2211-1G
TCA2211-1GEA 150.96 USD
TCA2211-1G TCA2211-5G
8-Aminoquinoline-N-oxide ≥98.0% (by GC, titration analysis)
8-Aminoquinoline-N-oxide

CAS Number: 92339-84-9
Molecular Formula: C9H8N2O
Molecular Weight: 160.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 110

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 110 °C
CAS Number: 92339-84-9
https://us.vwr.com/stibo/web/std.lang.all/04/01/28930401.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR