2,6-Dichlorobenzal chloride ≥98.0%

Danger

Synonyms: alpha,alpha,2,6-Tetrachlorotoluene2,6-Dichlorobenzal chloride2,6,α,α-Tetrachlorotolueneα,α,2,6-Tetrachlorotoluene

T0073-25G
TCT0073-025GEA 36.05 USD
TCT0073-025G
2,6-Dichlorobenzal chloride ≥98.0%
2,6-Dichlorobenzal chloride

CAS Number: 81-19-6
MDL Number: MFCD00016349
Molecular Formula: C7H4Cl4
Molecular Weight: 229.91
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 250
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 2,6-Dichlorobenzal Chloride

Formula: C₇H₄Cl₄
MW: 229.92 g/mol
Boiling Pt: 250 °C
Density: 1.53 (20 °C)
MDL Number: MFCD00016349
CAS Number: 81-19-6
EINECS: 201-332-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/44/11/28994411.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR