Tetrachlorobisphenol A ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2-Bis(3,5-dichloro-4-hydroxyphenyl)propane4,4'-Isopropylidenebis(2,6-dichlorophenol)

T0062-25G
TCT0062-025GEA 79.14 USD
TCT0062-025G
Tetrachlorobisphenol A ≥98.0% (by GC, titration analysis)
Tetrachlorobisphenol A

CAS Number: 79-95-8
MDL Number: MFCD00002178
Molecular Formula: C15H12Cl4O2
Molecular Weight: 366.06
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 134


Other Product Names / Synonyms: 2,2-Bis(3,5-dichloro-4-hydroxyphenyl)propane, 4,4'-Isopropylidenebis(2,6-dichlorophenol)

https://us.vwr.com/stibo/web/std.lang.all/44/04/28994404.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR