9,10,12,13-Tetrabromostearic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
9,10,12,13-Tetrabromooctadecanoic Acid
T0047-10G
TCT0047-010GEA
299.29
USD
TCT0047-010G
9,10,12,13-Tetrabromostearic acid ≥97.0% (by titrimetric analysis)
9,10,12,13-Tetrabromostearic acid
CAS Number: 1794-89-4
MDL Number: MFCD00021796
Molecular Formula: C18H32Br4O2
Molecular Weight: 600.07
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 115
Other Product Names / Synonyms: 9,10,12,13-Tetrabromooctadecanoic Acid
Formula:
C₁₈H₃₂Br₄O₂ MW: 600.07 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00021796 CAS Number: 1794-89-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...