Calcium-DL-tartrate tetrahydrate ≥98.0% (by titrimetric analysis)

Synonyms: DL-Tartaric Acid Calcium SaltRacemic Acid Calcium Salt

T0002-25G
TCT0002-025GEA 392.43 USD
TCT0002-025G
Calcium-DL-tartrate tetrahydrate ≥98.0% (by titrimetric analysis)
Calcium-DL-tartrate tetrahydrate

MDL Number: MFCD00151484
Molecular Formula: C4H6O6
Molecular Weight: 188.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: DL-Tartaric Acid Calcium Salt, Racemic Acid Calcium Salt

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR