5-Sulfoanthranilic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-5-sulfobenzoic acid2-Amino-5-sulphobenzoic acid

S0802-5G S0802-25G
TCS0802-25GEA 304.36 USD
TCS0802-25G TCS0802-5G
5-Sulfoanthranilic acid ≥96.0% (by HPLC, titration analysis)
5-Sulfoanthranilic acid

CAS Number: 3577-63-7
MDL Number: MFCD00035763
Molecular Formula: C7H7NO5S
Molecular Weight: 217.20
Purity/Analysis Method: 96.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 317


Other Product Names / Synonyms: 2-Amino-5-sulfobenzoic Acid

Formula: C₇H₇NO₅S
MW: 217.20 g/mol
Melting Pt: 317 °C
MDL Number: MFCD00035763
CAS Number: 3577-63-7
https://us.vwr.com/stibo/web/std.lang.all/42/11/28994211.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR