Sulfamethoxypyridazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide

S0591-5G S0591-25G
TCS0591-5GEA 35.5 USD
TCS0591-5G TCS0591-25G
Sulfamethoxypyridazine ≥98.0% (by HPLC, titration analysis)
Sulphamethoxypyridazine

CAS Number: 80-35-3
MDL Number: MFCD00057372
Molecular Formula: C11H12N4O3S
Molecular Weight: 280.30
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 182


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4-Amino-N-(6-methoxy-3-pyridazinyl)benzenesulfonamide, N-(6-Methoxy-3-pyridazinyl)sulfanilamide

Formula: C₁₁H₁₂N₄O₃S
MW: 280.3 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00057372
CAS Number: 80-35-3
https://us.vwr.com/stibo/web/std.lang.all/42/02/28994202.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR