Monosodium 2-sulfoterephthalate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Monosodium 2-sulphoterephthalate2-sulfoterephthalic acid Monosodium salt2-Sulphoterephthalic acid Monosodium saltMonosodium 2-Sulfoterephthalate

S0419-25G
TCS0419-025GEA 200.72 USD
TCS0419-025G
Monosodium 2-sulfoterephthalate ≥98.0% (by HPLC, titration analysis)
Monosodium 2-sulfoterephthalate

CAS Number: 19089-60-2
MDL Number: MFCD00059843
Molecular Formula: C8H6O7S
Molecular Weight: 268.17
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-Sulfoterephthalic Acid Monosodium Salt

Formula: C₈H₆O₇S
MW: 268.17 g/mol
MDL Number: MFCD00059843
CAS Number: 19089-60-2
https://us.vwr.com/stibo/web/std.lang.all/41/09/28994109.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR