1-Acenaphthenone ≥98.0%

Supplier: TCI America

Synonyms: Acenaphthylen-1(2H)-one

A2185-1G
TCA2185-1GEA 120.74 USD
TCA2185-1G
1-Acenaphthenone ≥98.0%
1-Acenaphthenone

CAS Number: 2235-15-6
MDL Number: MFCD00156651
Molecular Formula: C12H8O
Molecular Weight: 168.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 122

Formula: C₁₂H₈O
MW: 168.20 g/mol
Boiling Pt: 180 °C
Melting Pt: 122 °C
MDL Number: MFCD00156651
CAS Number: 2235-15-6
https://us.vwr.com/stibo/web/std.lang.all/03/74/28930374.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR