Acid red 9
Supplier: TCI America
Synonyms:
Silk Scarlet
S0140-25G
TCS0140-025GEA
60.88
USD
TCS0140-025G
Acid red 9
Acid red 9
CAS Number: 8003-59-6
MDL Number: MFCD00151319
Molecular Formula: C20H14N2O4S
Molecular Weight: 400.38
Form: Crystal
Color: Reddish Yellow
Other Product Names / Synonyms: Silk Scarlet
Formula:
C₂₀H₁₄N₂O₄S MW: 400.38 g/mol |
MDL Number:
MFCD00151319 CAS Number: 8003-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...