2-Sulfobenzaldehyde sodium salt ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Sodium 2-formylbenzenesulfonate
, Sodium 2-formylbenzenesulphonate
, Sodium 2-Formylbenzenesulfonate
, Sodium 2-Formylbenzenesulphonate
, 2-Formylbenzenesulfonic acid sodium salt
, 2-Formylbenzenesulphonic acid sodium salt
S0122-500G
S0122-25G
TCS0122-025GEA
73.3
USD
TCS0122-025G
TCS0122-500G
2-Sulfobenzaldehyde sodium salt ≥98.0% (by HPLC, titration analysis)
2-Sulfobenzaldehyde sodium salt
CAS Number: 1008-72-6
MDL Number: MFCD00007478
Molecular Formula: C7H6O4S
Molecular Weight: 208.16
Purity/Analysis Method:
Form: Crystal
Color: White
Other Product Names / Synonyms: Sodium 2-Formylbenzenesulfonate, 2-Formylbenzenesulfonic Acid Sodium Salt
Formula:
C₇H₆O₄S MW: 208.16 g/mol |
MDL Number:
MFCD00007478 CAS Number: 1008-72-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...