Strychnine hemisulfate pentahydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
S0094-25G
TCS0094-025GEA
226.48
USD
TCS0094-025G
Strychnine hemisulfate pentahydrate ≥97.0% (by titrimetric analysis)
Strychnine hemisulphate pentahydrate
CAS Number: 60491-10-3
MDL Number: MFCD00068328
Molecular Formula: C21H22N2O2
Molecular Weight: 766.91
Purity/Analysis Method:
Form: Crystal
Color: White
Formula:
C₄₂H₄₆N₄O₈S·5H₂O |
MDL Number:
MFCD00068328 CAS Number: 60491-10-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...