5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide

A2169-25G
TCA2169-25GEA 323 USD
TCA2169-25G
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97.0% (by total nitrogen basis)
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide

CAS Number: 31127-80-7
MDL Number: MFCD08063354
Molecular Formula: C16H20I3N3O7
Molecular Weight: 747.06
Purity/Analysis Method: 97.0% (N)
Form: Crystal
Melting point (°C): 275


Other Product Names / Synonyms: 5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide

Formula: C₁₆H₂₀I₃N₃O₇
MW: 747.06 g/mol
Melting Pt: 275 °C
MDL Number: MFCD08063354
CAS Number: 31127-80-7
https://us.vwr.com/stibo/web/std.lang.all/03/64/28930364.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR