Serotonin creatinine sulfate monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Hydroxytryptamine Creatinine Sulfate

S0036-1G
TCS0036-001GEA 72.47 USD
TCS0036-001G
Serotonin creatinine sulfate monohydrate ≥98.0% (by titrimetric analysis)
Serotonin creatinine sulfate monohydrate

CAS Number: 61-47-2
MDL Number: MFCD00149653
Molecular Formula: C10H12N2O
Molecular Weight: 387.41
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 215
Lambda max.: 273 nm (H2O)


Product Categories: Bioscience, Chemical Mediators
Other Product Names / Synonyms: 5-Hydroxytryptamine Creatinine Sulfate

Formula: C₁₀H₁₂N₂O
MW: 387.41 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00149653
CAS Number: 61-47-2
https://us.vwr.com/stibo/web/std.lang.all/39/10/28993910.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR