ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]) ≥98.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

Synonyms: Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate)2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt

A2166-1G
TCA2166-1GEA 79.2 USD
TCA2166-1G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]) ≥98.0% (by total nitrogen basis) for biochemical research
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

[for Biochemical Research]
CAS Number: 30931-67-0
MDL Number: MFCD00010404
Molecular Formula: C18H24N6O6S4
Molecular Weight: 548.67
Purity/Analysis Method: 98.0% (N)
Form: Crystal


Product Categories: Bioscience, Peroxidase Substrates, Redox Research Reagents
Other Product Names / Synonyms: 2,2'-Azinobis(3-ethylbenzothiazoline-6-sulfonic Acid Ammonium Salt)

Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
MDL Number: MFCD00010404
CAS Number: 30931-67-0
https://us.vwr.com/stibo/web/std.lang.all/03/60/28930360.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR