Rhodanine-N-propionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-(4-Oxo-2-thioxothiazolidin-3-yl)propanoic acid

R0060-5G R0060-25G
TCR0060-025GEA 464.87 USD
TCR0060-025G TCR0060-5G
Rhodanine-N-propionic acid ≥98.0% (by titrimetric analysis)
Rhodanine-N-propionic acid

CAS Number: 7025-19-6
MDL Number: MFCD00022558
Molecular Formula: C6H7NO3S2
Molecular Weight: 205.25
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: N-(2-Carboxyethyl)rhodanine

Formula: C₆H₇NO₃S₂
MW: 205.26 g/mol
MDL Number: MFCD00022558
CAS Number: 7025-19-6
EINECS: 230-307-7
https://us.vwr.com/stibo/web/std.lang.all/37/66/28993766.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR