Resorcinol monoacetate 50.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Acetic acid 3-Hydroxyphenyl ester
, 3-Hydroxyphenyl acetate
R0009-500G
R0009-25G
TCR0009-500GEA
233
USD
TCR0009-500G
TCR0009-025G
Resorcinol monoacetate 50.0% (by GC)
Resorcinol monoacetate
CAS Number: 102-29-4
MDL Number: MFCD00002266
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method:
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 283
Flash Point (°C): 110
Specific Gravity (20/20): 1.20
Other Product Names / Synonyms: Acetic Acid 3-Hydroxyphenyl Ester, 3-Hydroxyphenyl Acetate
Formula:
C₈H₈O₃ Boiling Pt: 283 °C Density: 1.20 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002266 UN: 2810 ADR: 6.1,III |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...