Quinizarin green ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Solvent Green 3D and C Green No. 61,4-Bis(p-tolylamino)anthracene-9,10-dione1,4-Bis(p-toluidino)-9,10-anthraquinone

Q0021-25G
TCQ0021-025GEA 130.61 USD
TCQ0021-025G
Quinizarin green ≥98.0% (by total nitrogen basis)
1,4-Bis(p-toluidino)-9,10-anthraquinone

CAS Number: 128-80-3
MDL Number: MFCD00001195
Molecular Formula: C28H22N2O2
Molecular Weight: 418.50
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: Purple
Lambda max.: 638 nm (Acetone)


Other Product Names / Synonyms: Solvent Green 3

Formula: C₂₈H₂₂N₂O₂
MW: 418.496 g/mol
MDL Number: MFCD00001195
CAS Number: 128-80-3
https://us.vwr.com/stibo/web/std.lang.all/36/88/28993688.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR