[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel ≥96.0% (by titrimetric analysis)

Supplier: TCI America

P1738-100MG
TCP1738-100MGEA 40 USD
TCP1738-100MG
[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel ≥96.0% (by titrimetric analysis)
[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel

CAS Number: 264921-97-3
Molecular Formula: C16H13N3NiO3
Molecular Weight: 353.99
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Color: Pale Yellow

Formula: C₁₆H₁₃N₃NiO₃
CAS Number: 264921-97-3
https://us.vwr.com/stibo/web/std.lang.all/30/58/28993058.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR