D-Psicose ≥99.0% (by HPLC)
Supplier: TCI America
Synonyms:
(3R,4R,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one
P1699-1G
P1699-100MG
TCP1699-1GEA
504.16
USD
TCP1699-1G
TCP1699-100MG
D-Psicose ≥99.0% (by HPLC)
D-Psicose
CAS Number: 551-68-8
MDL Number: MFCD00083478
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 109
Specific rotation [a]20/D: 4 deg (C=1, H2O)
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 109 °C |
MDL Number:
MFCD00083478 CAS Number: 551-68-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...