(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran, Tetrahydrofuran Adduct ≥90.0% (by titrimetric analysis)

Synonyms: Bis(tricyclohexylphosphine)-3-phenyl-1H-inden-1-ylideneruthenium(II) dichloride

P1688-100MG
TCP1688-100MGEA 66.95 USD
TCP1688-100MG
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran, Tetrahydrofuran Adduct ≥90.0% (by titrimetric analysis)
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran

CAS Number: 250220-36-1
MDL Number: MFCD07369034
Molecular Formula: C51H76Cl2P2Ru
Molecular Weight: 923.09
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: Deep Yellow

Formula: C₅₁H₇₆Cl₂P₂Ru
MW: 923.09 g/mol
MDL Number: MFCD07369034
CAS Number: 250220-36-1
https://us.vwr.com/stibo/web/std.lang.all/30/27/28993027.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR