(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene ≥95.0%

A2128-5G A2128-1G
TCA2128-1GEA 181.28 USD
TCA2128-1G TCA2128-5G
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene ≥95.0%
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene

CAS Number: 911200-98-1
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 74
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: -104 deg (C=1, MeOH)

Formula: C₁₀H₁₄O
Density: 0.98 (20 °C)
Flash Pt: 74 °C
CAS Number: 911200-98-1
https://us.vwr.com/stibo/web/std.lang.all/02/99/28930299.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR