trans-4-(4-Propylcyclohexyl)benzonitrile ≥98.0%

Supplier: TCI America

Synonyms: 4-(trans-4-propylcyclohexyl)benzonitrile

P1617-5G
TCP1617-5GEA 113 USD
TCP1617-5G
trans-4-(4-Propylcyclohexyl)benzonitrile ≥98.0%
trans-4-(4-Propylcyclohexyl)benzonitrile

CAS Number: 61203-99-4
MDL Number: MFCD04038799
Molecular Formula: C16H21N
Molecular Weight: 227.35
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 45


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 1-Cyano-4-(trans-4-propylcyclohexyl)benzene

Formula: C₁₆H₂₁N
MW: 227.3446 g/mol
Melting Pt: 45 °C
MDL Number: MFCD04038799
CAS Number: 61203-99-4
https://us.vwr.com/stibo/web/std.lang.all/21/65/28992165.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR