3A-Amino-3A-deoxy-(2aS,3aS)-γ-cyclodextrin hydrate ≥95.0% (by HPLC)

A2123-200MG
TCA2123-200MGEA 99.9 USD
TCA2123-200MG
3A-Amino-3A-deoxy-(2aS,3aS)-γ-cyclodextrin hydrate ≥95.0% (by HPLC)
3A-Amino-3A-deoxy-(2aS,3aS)-gamma-cyclodextrin hydrate

CAS Number: 189307-64-0
Molecular Formula: C48H81NO39
Molecular Weight: 1296.14
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 165 deg (C=1, H2O)

Formula: C₄₈H₈₁NO₃₉
CAS Number: 189307-64-0
https://us.vwr.com/stibo/web/std.lang.all/02/92/28930292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR