Ammonium perfluorooctanoate ≥98.0% (by titrimetric analysis)

P1449-25G
TCP1449-25GEA 133.17 USD
TCP1449-25G
Ammonium perfluorooctanoate ≥98.0% (by titrimetric analysis)
Ammonium perfluorooctanoate

CAS Number: 3825-26-1
MDL Number: MFCD00042599
Molecular Formula: C8H4F15NO2
Molecular Weight: 431.10
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Ammonium Perfluorooctanoate, Pentadecafluorooctanoic Acid Ammonium Salt, Perfluorooctanoic Acid Ammonium Salt

Formula: C₈H₄F₁₅NO₂
MDL Number: MFCD00042599
CAS Number: 3825-26-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR