3-Phenoxypropiononitrile ≥98.0%
Supplier: TCI America
P1426-5G
TCP1426-5GEA
53.8
USD
TCP1426-5G
3-Phenoxypropiononitrile ≥98.0%
3-Phenoxypropiononitrile
CAS Number: 3055-86-5
MDL Number: MFCD00013822
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 120
Melting point (°C): 60
Formula:
C₉H₉NO Boiling Pt: 120 °C Melting Pt: 60 °C |
MDL Number:
MFCD00013822 CAS Number: 3055-86-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...