Methyl pentafluorobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl Pentafluorobenzoate

P1418-25G P1418-5G
TCP1418-25GEA 66 USD
TCP1418-25G TCP1418-5G
Methyl pentafluorobenzoate ≥98.0%
Methyl pentafluorobenzoate

CAS Number: 36629-42-2
MDL Number: MFCD00012172
Molecular Formula: C8H3F5O2
Molecular Weight: 226.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 78
Specific Gravity (20/20): 1.54


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Pentafluorobenzoic Acid Methyl Ester, Methyl Perfluorobenzoate, Perfluorobenzoic Acid Methyl Ester

Formula: C₈H₃F₅O₂
MW: 226.1002 g/mol
Density: 1.54 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00012172
CAS Number: 36629-42-2
https://us.vwr.com/stibo/web/std.lang.all/20/77/28992077.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR