Poly[4-(diacetoxyiodo)styrene]
Supplier: TCI America
Synonyms:
PSDIB
, Polystyrene Iodosoacetate
P1415-5G
P1415-1G
TCP1415-1GEA
204.94
USD
TCP1415-1G
TCP1415-5G
Poly[4-(diacetoxyiodo)styrene]
Poly[4-(diacetoxyiodo)styrene]
CAS Number: 36290-94-5
MDL Number: MFCD06798811
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: Polystyrene Iodosoacetate, PSDIB
MDL Number:
MFCD06798811 CAS Number: 36290-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...