(S,S)-(+)-2,4-Pentanediol ≥98.0%
Supplier: TCI America
Synonyms:
(S,S)-(+)-2,4-Dihydroxy pentane
, (2S,4S)-(+)-Pentanediol
, (2S,4S)-(+)-2,4-Pentanediol
, (2S,4S)-2,4-Pentanediol
CAS Number: 72345-23-4
MDL Number: MFCD00064276
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 48
Flash Point (°C): 101
Specific rotation [a]20/D: 54 deg (C=10, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,4S)-(+)-2,4-Dihydroxypentane
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Melting Pt: 48 °C Flash Pt: 101 °C |
MDL Number:
MFCD00064276 CAS Number: 72345-23-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...