5-Phenyl-2-furancarboxylic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Phenyl-2-furoic Acid

P1394-1G
TCP1394-1GEA 133 USD
TCP1394-1G
5-Phenyl-2-furancarboxylic acid ≥96.0% (by titrimetric analysis)
5-Phenyl-2-furancarboxylic acid

CAS Number: 52938-97-3
MDL Number: MFCD00456304
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 151


Other Product Names / Synonyms: 5-Phenyl-2-furoic Acid

Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00456304
CAS Number: 52938-97-3
https://us.vwr.com/stibo/web/std.lang.all/20/38/28992038.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR