Artemisinin ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
Qinghaosu
, Arteannuin
A2118-1G
A2118-5G
TCA2118-5GEA
270.56
USD
TCA2118-5G
76218-738
Artemisinin ≥97.0% (by HPLC)
Artemisinin
CAS Number: 63968-64-9
MDL Number: MFCD00081057
Molecular Formula: C15H22O5
Molecular Weight: 282.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 75 deg (C=0.5, MeOH)
Product Categories: Bioscience, Antiprotozoal Ingredients, Sesquiterpenes
Other Product Names / Synonyms: Arteannuin, Qinghaosu
Formula:
C₁₅H₂₂O₅ MW: 282.34 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00081057 CAS Number: 63968-64-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...