1-Propylpyridinium chloride
Supplier: TCI America
P1393-5G
TCP1393-5GEA
282.05
USD
TCP1393-5G
1-Propylpyridinium chloride
1-Propylpyridinium chloride
CAS Number: 23271-47-8
MDL Number: MFCD06797153
Molecular Formula: C8H12ClN
Molecular Weight: 157.64
Form: Crystal
Color: Pale Yellow
Formula:
C₈H₁₂ClN |
MDL Number:
MFCD06797153 CAS Number: 23271-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...