(1R,2S)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥98.0%
Supplier: TCI America
Synonyms:
(1R,2S)-N,N-Tetramethylenenorephedrine
P1374-1G
P1374-5G
TCP1374-001GEA
86.58
USD
TCP1374-001G
TCP1374-5G
(1R,2S)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥98.0%
(1R,2S)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol
CAS Number: 127641-25-2
MDL Number: MFCD01632702
Molecular Formula: C13H19NO
Molecular Weight: 205.30
Purity/Analysis Method:
Form: Crystal
Color: White
Specific rotation [a]20/D: 14 deg (C=2, CHCl3)
Other Product Names / Synonyms: (1R,2S)-N,N-Tetramethylenenorephedrine
Formula:
C₁₃H₁₉NO MW: 205.30 g/mol |
MDL Number:
MFCD01632702 CAS Number: 127641-25-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...