(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol(S)-2,2,2-Trifluoro-1-phenylethanol

P1368-1G
TCP1368-001GEA 296 USD
TCP1368-001G
(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol ≥98.0%
(S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol

CAS Number: 340-06-7
MDL Number: MFCD00077845
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 127
Flash Point (°C): 84
Specific Gravity (20/20): 1.30
Specific rotation [a]20/D: 31.5 deg (neat)


Other Product Names / Synonyms: (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 127 °C
Density: 1.30 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00077845
CAS Number: 340-06-7
https://us.vwr.com/stibo/web/std.lang.all/20/15/28992015.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR