(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol

Supplier: TCI America

Synonyms: (R)-(-)-1-Phenyl-2,2,2-trifluoroethanol

P1367-1G
TCP1367-001GEA 367 USD
TCP1367-001G
(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol
(R)-(-)-alpha-(Trifluoromethyl)benzyl alcohol

CAS Number: 10531-50-7
MDL Number: MFCD00077844
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 99
Flash Point (°C): 87
Specific Gravity (20/20): 1.30
Specific rotation [a]20/D: -31.5 deg (neat)


Other Product Names / Synonyms: (R)-(-)-1-Phenyl-2,2,2-trifluoroethanol

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 99 °C
Density: 1.30 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00077844
CAS Number: 10531-50-7
https://us.vwr.com/stibo/web/std.lang.all/20/13/28992013.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR